Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCER1G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109716 Supplier Invitrogen™ Supplier No. PA5109716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGEPQLCYIL DAILFLYGIV LTLLYCRLKI QVRKAAITSY EKSDGVYTGL STRNQETYET LKHEKPPQ.

The high affinity IgE receptor is a key molecule involved in allergic reactions. It is a tetramer composed of 1 alpha, 1 beta, and 2 gamma chains. The gamma chains are also subunits of other Fc receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCER1G
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FCER1G
Gene Accession No. P20411, P20491
Gene Alias AI573376; CD23; Fc epsilon receptor Ig; Fc fragment of IgE receptor Ig; Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide; Fc receptor gamma-chain; Fc receptor, IgE, high affinity I, gamma polypeptide; Fce1g; fc-epsilon RI-gamma; FcepsilonRI; FCER1G; FceRI gamma; FcR[g]; FCRG; FcRgamma; FcR-gamma; gamma polypeptide; high affinity immunoglobulin epsilon receptor subunit gamma; IgE Fc receptor subunit gamma; immunoglobulin E receptor, high affinity, gamma chain; Ly-50
Gene Symbols FCER1G
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 19-86 of human FCER1G (NP_004097.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14127, 25441
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.