Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCER1G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528832 Supplier Invitrogen™ Supplier No. PA528832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Fc epsilonRIgamma tansfected 293T cell. Predicted reactivity: Rat (100%), Dog (100%), Pig (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The high affinity IgE receptor is a key molecule involved in allergic reactions. It is a tetramer composed of 1 alpha, 1 beta, and 2 gamma chains. The gamma chains are also subunits of other Fc receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCER1G
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene FCER1G
Gene Accession No. P20491, P30273
Gene Alias AI573376; CD23; Fc epsilon receptor Ig; Fc fragment of IgE receptor Ig; Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide; Fc receptor gamma-chain; Fc receptor, IgE, high affinity I, gamma polypeptide; Fce1g; fc-epsilon RI-gamma; FcepsilonRI; FCER1G; FceRI gamma; FcR[g]; FCRG; FcRgamma; FcR-gamma; gamma polypeptide; high affinity immunoglobulin epsilon receptor subunit gamma; IgE Fc receptor subunit gamma; immunoglobulin E receptor, high affinity, gamma chain; Ly-50
Gene Symbols FCER1G
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Intracellular domain of human FCER1G. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14127, 2207
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.