Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCER1A Mouse anti-Human, Clone:1F2A9, Abnova™

Catalog No. 89334525 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-525 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-525 Supplier Abnova Corporation Supplier No. MAB17840
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human FCER1A.

The IgE receptor plays a central role in allergic disease, coupling allergen and mast cell to initiate the inflammatory and immediate hypersensitivity responses that are characteristic of disorders such as hay fever and asthma. The allergic response occurs when 2 or more high-affinity IgE receptors are crosslinked via IgE molecules that in turn are bound to an allergen (antigen) molecule. A perturbation occurs that brings about the release of histamine and proteases from the granules in the cytoplasm of the mast cell and leads to the synthesis of prostaglandins and leukotrienes--potent effectors of the hypersensitivity response. The IgE receptor consists of 3 subunits: alpha, beta (MIM 147138), and gamma (MIM 147139); only the alpha subunit is glycosylated.[supplied by OMIM]

Specifications

Antigen FCER1A
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1F2A9
Conjugate Unconjugated
Description Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene FCER1A
Gene Alias FCE1A, FcERI
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 42-103 of human FCER1A from E. coli.
Quantity 100 μg
Gene ID (Entrez) 2205
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.