Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCER1G Recombinant Rabbit Monoclonal Antibody (HL1418)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547037
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The high affinity IgE receptor is a key molecule involved in allergic reactions. It is a tetramer composed of 1 alpha, 1 beta, and 2 gamma chains. The gamma chains are also subunits of other Fc receptors.
Specifications
FCER1G | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P20411, P20491, P30273 | |
FCER1G | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FCER1G. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
HL1418 | |
Unconjugated | |
FCER1G | |
AI573376; CD23; Fc epsilon receptor Ig; Fc fragment of IgE receptor Ig; Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide; Fc receptor gamma-chain; Fc receptor, IgE, high affinity I, gamma polypeptide; Fce1g; fc-epsilon RI-gamma; FcepsilonRI; FCER1G; FceRI gamma; FcR[g]; FCRG; FcRgamma; FcR-gamma; gamma polypeptide; high affinity immunoglobulin epsilon receptor subunit gamma; IgE Fc receptor subunit gamma; immunoglobulin E receptor, high affinity, gamma chain; Ly-50 | |
Rabbit | |
Protein A | |
RUO | |
14127, 2207, 25441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction