Learn More
FCER2 Mouse anti-Human, Clone: OTI6C2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801554

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.Specifications
FCER2 | |
Monoclonal | |
Unconjugated | |
FCER2 | |
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF | |
Mouse | |
Affinity Chromatography | |
RUO | |
2208 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6C2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P06734 | |
FCER2 | |
Human recombit protein fragment corresponding to amino acids 48-321 of human FCER2 produced in SF9 Cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.