Learn More
Promega Corporation FcgammaRI ADCP Reporter Bioassay, Core Kit
Catalog No. PRGA1341 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Type:
Bioassay
Catalog No. Type
PRGA1341 Bioassay
1 options
Catalog No. PRGA1341 Supplier Promega Corporation Supplier No. GA1341
Measure potency and stability of biologics that bind and activate ADCP through FcγRI is prequalified according to ICH guidelines. No primary cell culture is required. Supplied in convenient thaw-and-use format, also available in cell propagation model (CPM) format.
  • Measure potency and stability of biologics that bind and activate ADCP through FcγRI.
  • Prequalified according to ICH guidelines.
  • No primary cell culture required.
  • Supplied in convenient thaw-and-use format.
  • Also available in cell propagation model (CPM) format.
  • High sensitivity and specificity.
  • Consistent and reproducible results.
  • Suitable for research and clinical applications.
  • Streamlined workflow reduces hands-on time.
  • Reliable and efficient solution for ADCP bioassays.
Specifications
Detection Method Bioluminescence
Type Bioassay
Cell Line Jurkat
Druggable Target Nuclear Receptors