Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCGBP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$646.00
Specifications
Antigen | FCGBP |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FCGBP Polyclonal specifically detects FCGBP in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FCGBP | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
8857 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GHRFDFQGTCEYLLSAPCHGPPLGAENFTVTVANEHRGSQAVSYTRSVTLQIYNHSLTLSARWPRKLQVDGVFVALPFQLDSLLHAHLSGADVVVTTTSGLSLAFDGDSF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
Fc fragment of IgG binding protein, FC(GAMMA)BP, fcgamma-binding protein antigen, fcgammaBP, Human Fc gamma BP, IgG Fc binding protein, IgGFc-binding protein | |
FCGBP | |
IgG | |
Affinity Purified | |
Specificity of human FCGBP antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title