Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCGR1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526205
Description
FCGR1B Polyclonal Antibody for Western Blot, IHC (P), Flow
FCGR1B may bind to the Fc region of immunoglobulins gamma with a low affinity compared to FCGR1A. This protein may function in the humoral immune response.
Specifications
FCGR1B | |
Polyclonal | |
Unconjugated | |
FCGR1B | |
CD64b; Fc fragment of IgG receptor Ib; Fc fragment of IgG, high affinity Ib, receptor (CD64); Fc gamma receptor Ib; Fc-gamma receptor I B2; fc-gamma RIB; FCGR1B; FcRIB; hFcgammaRIB; high affinity immunoglobulin gamma Fc receptor IB; IGFRB; IgG Fc receptor IB | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2210 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q92637 | |
FCGR1B | |
KLH conjugated synthetic peptide between 248-276 amino acids from the C-terminal region of human FCGR1B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction