Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCGR2A / CD32a Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | FCGR2A / CD32a |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0361
![]()
|
Proteintech Group Inc
156251AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Specifications
| FCGR2A / CD32a | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P12318 | |
| 2212 | |
| FCGR2A / CD32a Fusion Protein Ag8072 | |
| Primary | |
| -20°C | |
| FCGR2A |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD32, CD32A, CDw32, Fc gamma RII a, Fc gamma RIIa, FCG2, FcGR, FCGR2, FCGR2A, FCGR2A1, FcRII a, IGFR2, IgG Fc receptor II a | |
| FCGR2A | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title