Learn More
Invitrogen™ FCGR2A Recombinant Rabbit Monoclonal Antibody (3B6)

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | FCGR2A |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 3B6 |
| Concentration | 0.65 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | FCGR2A |
| Gene Accession No. | P12318 |
| Gene Alias | CD16; CD32; CD32A; CDw32; Fc fragment of IgG low affinity IIa receptor for (CD32); Fc fragment of IgG receptor IIa; Fc fragment of IgG, low affinity IIa, receptor; Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc gamma receptor IIa; Fc receptor, IgG, low affinity III; FCG2; fc-gamma RII-a; Fc-gamma RIII; fc-gamma-RIIa; FcGR; FCGR2; FCGR2A; FCGR2A1; Fcgr3; Fcgr3a; fcRII-a; fcRIII; FGCR; IGFR2; IgG Fc receptor II-a; igG Fc receptor III; Immunoglobulin G Fc receptor II; low affinity immunoglobulin gamma Fc region receptor II-a; Low affinity immunoglobulin gamma Fc region receptor III |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.