Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR2A Mouse anti-Human, Clone: 13D7, Abnova™

Catalog No. 89934130 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-130 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-130 Supplier Abnova Corporation Supplier No. MAB12859
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human FCGR2A.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Specifications

Antigen FCGR2A
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 13D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human FCGR2A.
Dilution Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
Gene Alias CD32/CD32A/CDw32/FCG2/FCGR2/FCGR2A1/FcGR/IGFR2/MGC23887/MGC30032
Gene Symbols FCGR2A
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human FCGR2A.
Purification Method Protein A/G purification
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2212
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.