Learn More
FCGR2A Mouse anti-Human, Clone: OTI13G9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500646

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Specifications
FCGR2A | |
Monoclonal | |
Unconjugated | |
FCGR2A | |
CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2212 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI13G9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P12318 | |
FCGR2A | |
Full length human recombit protein of human FCGR2A produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.