Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCGR2A Mouse anti-Human, Clone: OTI16D6, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500667

Description
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Specifications
FCGR2A | |
Monoclonal | |
Unconjugated | |
FCGR2A | |
CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2212 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
OTI16D6 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P12318 | |
FCGR2A | |
Full length human recombit protein of human FCGR2A produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction