Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCGR2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116569 Supplier Invitrogen™ Supplier No. PA5116569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: K-562, Mouse spleen Immunogen sequence: QAAAPPKAVL KLEPPWINVL QEDSVTLTCQ GARSPESDSI QWFHNGNLIP THTQPSYRFK ANNNDSGEYT CQTGQTSLSD PVHLTVLSEW LVLQTPHLEF QEGETIMLRC HSWKDKPLVK VTFFQNGKSQ KFSHLDPTFS IPQANHSHSG DYHCTGNIGY TLFSSKPVTI TVQVPSMGSS SPMGIIVAVV IATAVAAIVA AVVALIYCRK KRISANSTDP VKAAQFEPPG RQMIAIRKRQ LEETNNDYET ADGGYMTLNP RAPTDDDKNI YLTLPPNDHV NSNN.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCGR2A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene FCGR2A
Gene Accession No. P12318
Gene Alias CD16; CD32; CD32A; CDw32; Fc fragment of IgG low affinity IIa receptor for (CD32); Fc fragment of IgG receptor IIa; Fc fragment of IgG, low affinity IIa, receptor; Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc gamma receptor IIa; Fc receptor, IgG, low affinity III; FCG2; fc-gamma RII-a; Fc-gamma RIII; fc-gamma-RIIa; FcGR; FCGR2; FCGR2A; FCGR2A1; Fcgr3; Fcgr3a; fcRII-a; fcRIII; FGCR; IGFR2; IgG Fc receptor II-a; igG Fc receptor III; Immunoglobulin G Fc receptor II; low affinity immunoglobulin gamma Fc region receptor II-a; Low affinity immunoglobulin gamma Fc region receptor III
Gene Symbols FCGR2A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 34-316 of human FCGR2A (NP_0011296911).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116591, 2212
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.