Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCGR2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102409 Supplier Invitrogen™ Supplier No. PA5102409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CD32.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCGR2A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FCGR2A
Gene Accession No. P12318
Gene Alias CD16; CD32; CD32A; CDw32; Fc fragment of IgG low affinity IIa receptor for (CD32); Fc fragment of IgG receptor IIa; Fc fragment of IgG, low affinity IIa, receptor; Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc gamma receptor IIa; Fc receptor, IgG, low affinity III; FCG2; fc-gamma RII-a; Fc-gamma RIII; fc-gamma-RIIa; FcGR; FCGR2; FCGR2A; FCGR2A1; Fcgr3; Fcgr3a; fcRII-a; fcRIII; FGCR; IGFR2; IgG Fc receptor II-a; igG Fc receptor III; Immunoglobulin G Fc receptor II; low affinity immunoglobulin gamma Fc region receptor II-a; Low affinity immunoglobulin gamma Fc region receptor III
Gene Symbols FCGR2A
Host Species Rabbit
Immunogen A synthesized peptide derived from human FCGR2A(Accession P12318), corresponding to amino acid residues Q258-P308.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2212
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.