Learn More
Description
- Fc fragment of IgG, low affinity IIb, receptor (CD32)
- Transfected cell line: 293T
- Plasmid: pCMV-FCGR2B full-length
- Host: Human
- Theoretical molecular weight: 34.21kDa
- Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | FCGR2B 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | NM_004001 |
| Molecular Weight (g/mol) | 34.21 |
| pH Range | 6.8 |
| Plasmid | pCMV-FCGR2B full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-FCGR2B antibody (H00002213-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.