Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR2B, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89999249 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-249 Supplier Abnova Corporation Supplier No. PAB31633
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen FCGR2B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA (1:10000) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide).
Gene Fc fragment of IgG, low affinity IIb, receptor (CD32)
Gene Alias CD32/CD32B/FCG2/FCGR2/IGFR2
Gene Symbols FCGR2B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 230-310 of human FCGR2B (phospho Y292).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2213
Test Specificity This antibody detects endogenous levels of FCGR2B protein only when phosphorylated at Y292.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.