Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCGR2B Recombinant Rabbit Monoclonal Antibody (HL1950)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550351
Description
FCGR2B Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications
| FCGR2B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P31994 | |
| FCGR2B | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CD32B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1950 | |
| Unconjugated | |
| FCGR2B | |
| AI528646; CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG receptor IIb; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); fc gamma receptor IIB; Fc gamma RIIb; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; FC-gamma RII; fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma-RII; fc-gamma-RIIa; FcgammaRIIB; fc-gamma-RIIb; FcgammaRIIB2; FcGR; Fcgr2; Fcgr2a; FCGR2A1; FCGR2B; FcgRII; Fcr-2; Fcr-3; fcRII; fcRII-a; fcRII-b; FGFR2B; IGFR2; IgG Fc receptor II beta; igG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; LyM-1; Ly-m20; Lymphocyte antigen 17 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2213 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction