Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fcgr3, anti-Mouse, Clone: 2.4G2, Abnova™

Catalog No. 89032519 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-519 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-519 Supplier Abnova Corporation Supplier No. RAB00176
Add to Cart
Add to Cart

Monoclonal Antibody

Specifications

Antigen Fcgr3
Applications Blocking Assay, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2.4G2
Conjugate Unconjugated
Dilution Blocking Flow Cytometry (1 ug/mL) Immunofluorescence Immunohistochemistry (5 ug/mL) Immunoprecipitation (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% Proclin 300).
Gene Fc receptor, IgG, low affinity III
Gene Alias CD16
Gene Symbols Fcgr3
Immunogen Original antibody is raised against mouse BALB/c macrophage J774 cell line.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14131/14130
Test Specificity This antibody binds to an epitope on the extracellular domains of the mouse Fc gamma III and Fc gamma II receptors, thereby inhibiting their ability to bind antibodies via their Fc region. Also binding the Fc gamma I receptor (CD64) via its Fc domain.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.