Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR3A Mouse anti-Human, Clone: 3G8 (PerCP), Abnova™

Catalog No. 89033884 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-884 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-884 Supplier Abnova Corporation Supplier No. MAB13819
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human FCGR3A.

  • High specificity
  • High reactivity

Specifications

Antigen FCGR3A
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 3G8
Conjugate PerCP
Description Mouse monoclonal antibody raised against human FCGR3A.
Dilution Flow Cytometry (20 uL/106 cells) Functional Study Immunohistochemistry (Frozen sections) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Gene Accession No. P08637
Gene Alias CD16/CD16A/FCG3/FCGR3/FCGRIII/FCR-10/FCRIII/FCRIIIA/IGFR3
Gene Symbols FCGR3A
Host Species Mouse
Immunogen Human polymorphonuclear leukocytes.
Purification Method Protein A/G purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.