Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR3A Mouse anti-Human, Clone: CB16, Abnova™

Catalog No. 89034760 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-760 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-760 Supplier Abnova Corporation Supplier No. MAB14677

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human FCGR3A.

  • High specificity
  • High reactivity

Specifications

Antigen FCGR3A
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone CB16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human FCGR3A.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA and 0.05% azide).
Gene Alias CD16/CD16A/FCG3/FCGR3/FCGRIII/FCR-10/FCRIII/FCRIIIA/IGFR3
Gene Symbols FCGR3A
Host Species Mouse
Immunogen Human CD16
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity Recognizes CD16 (Fc gamma RIII), the low-affinity receptor for IgG, with an apparent molecular weight of 50-80kDa.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.