Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FcgR4/CD16-2 Antibody (012), PE , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP305890PE
Description
FcgR4/CD16-2 Monoclonal antibody specifically detects FcgR4/CD16-2 in Mouse samples. It is validated for Flow CytometrySpecifications
FcgR4/CD16-2 | |
Monoclonal | |
PE | |
Rabbit | |
Protein A purified | |
RUO | |
246256 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
012 | |
PBS | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FcgR4/CD16-2 (NP_653142.2; Met 1-Gln 203). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction