Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FcgR4/CD16-2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305900B
Description
FcgR4/CD16-2 Polyclonal specifically detects FcgR4/CD16-2 in Mouse samples. It is validated for ELISA.Specifications
FcgR4/CD16-2 | |
Polyclonal | |
ELISA | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
246256 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Biotin | |
PBS | |
Produced in rabbits immunized with purified, recombinant Mouse FcgR4/CD16-2 (NP_653142.2; Met 1-Gln 203;). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction