Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCHO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544284 Supplier Invitrogen™ Supplier No. PA544284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SPENVEDSGL DSPSHAAPGP SPDSWVPRPG TPQSPPSCRA PPPEARGIRA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

FCHO1 is involved in clathrin-coated vesicle formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCHO1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FCHO1
Gene Accession No. O14526
Gene Alias 3322402E17Rik; F-BAR domain only protein 1; FCH and mu domain containing endocytic adaptor 1; FCH domain only 1; FCH domain only protein 1; FCHO1; KIAA0290; N28152; unm_sa2424
Gene Symbols FCHO1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human FCHO1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23149
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.