Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCHO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566548 Supplier Invitrogen™ Supplier No. PA566548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGPQSVPLQLS AHWQCGATLT QVSVEYGYRP GATAVPTPLT NVQILLPVGE PVTNVRLQPA ATWNLEEKRL TW Highest antigen sequence identity to the following orthologs - mouse 86%, rat 85%.

FCHO1 is involved in clathrin-coated vesicle formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCHO1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FCHO1
Gene Accession No. O14526
Gene Alias 3322402E17Rik; F-BAR domain only protein 1; FCH and mu domain containing endocytic adaptor 1; FCH domain only 1; FCH domain only protein 1; FCHO1; KIAA0290; N28152; unm_sa2424
Gene Symbols FCHO1
Host Species Rabbit
Immunogen Recombinant Human FCHO1. Recombinant protein control fragment (Product #RP-105250).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23149
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.