Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCHO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583568
Description
Immunogen sequence: GAKAFRLPGL SRREREPEPP AAVDFLEPDS GTCPEVDEEG FTVRPDVTQN STAEPSRFSS SDSDFDDEEP RKFYVHIKPA.
FCHO1 is involved in clathrin-coated vesicle formation.
Specifications
FCHO1 | |
Polyclonal | |
Unconjugated | |
FCHO1 | |
3322402E17Rik; F-BAR domain only protein 1; FCH and mu domain containing endocytic adaptor 1; FCH domain only 1; FCH domain only protein 1; FCHO1; KIAA0290; N28152; unm_sa2424 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23149 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14526 | |
FCHO1 | |
Recombinant protein corresponding to Human FCHO1. Recombinant protein control fragment (Product #RP-99067). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction