Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCHO2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP236524AF532
Description
FCHO2 Polyclonal antibody specifically detects FCHO2 in Human samples. It is validated for Western BlotSpecifications
FCHO2 | |
Polyclonal | |
Western Blot | |
FCH domain only 2 | |
The immunogen recognized by this antibody maps to a region between residue 125 to 175 of human FCH Domain Only 2 using the numbering given in entry NP_620137.2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
115548 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction