Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCHO2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534390
Description
A suggested positive control is rat heart tissue lysate. PA5-34390 can be used with blocking peptide PEP-1433.
FCHO2 (FCH domain only 2) is a ubiquitously expressed member of the FCFBS superfamily characterized by FES-CIP4 homology (FCH) domain, an FBH domain, and an SH3 domain. Both FCHO2 and the related protein FCHO1 are mediators of clathrin-mediated endocytosis, acting to sculpt the initial bud site and recruit the clathrin machinery for clathrin-coated vesicle formation. FCHO2 binds to Eps15, an important adaptor protein in clathrin-mediated endocytosis.
Specifications
FCHO2 | |
Polyclonal | |
Unconjugated | |
FCHO2 | |
5832424M12Rik; AA387320; F-BAR domain only protein 2; FCH domain only 2; FCH domain only protein 2; FCHO2; RGD1565396 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115548, 218503, 309129 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
D3ZYR1, Q0JRZ9, Q3UQN2 | |
FCHO2 | |
A 15 amino acid synthetic peptide near the center of human FCHO2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction