Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCHO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558060
Description
Immunogen sequence: KAAVKSKKAT DTYKLYVEKY ALAKADFEQK MTETAQKFQD IEETHLIHIK EIIGSLSNAI KEIHLQIG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
FCHO2 (FCH domain only 2) is a ubiquitously expressed member of the FCFBS superfamily characterized by FES-CIP4 homology (FCH) domain, an FBH domain, and an SH3 domain. Both FCHO2 and the related protein FCHO1 are mediators of clathrin-mediated endocytosis, acting to sculpt the initial bud site and recruit the clathrin machinery for clathrin-coated vesicle formation. FCHO2 binds to Eps15, an important adaptor protein in clathrin-mediated endocytosis.
Specifications
FCHO2 | |
Polyclonal | |
Unconjugated | |
FCHO2 | |
5832424M12Rik; AA387320; F-BAR domain only protein 2; FCH domain only 2; FCH domain only protein 2; FCHO2; RGD1565396 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115548 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0JRZ9 | |
FCHO2 | |
Recombinant protein corresponding to Human FCHO2. Recombinant protein control fragment (Product #RP-96911). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction