Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCHSD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560514
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560514 Supplier Invitrogen™ Supplier No. PA560514
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHLDHYYQEE LPALLKALVS ELSEHLRDPL TSLSHTELEA AEVILEHAHR GEQTTSQVSW EQDLKLFLQE PGVFSPTPPQ QFQPAGTDQV CVLEWG Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

FCHSD1 gene ontology annotations related to this gene include neuromuscular synaptic transmission; regulation of actin filament polymerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCHSD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FCHSD1
Gene Accession No. Q86WN1
Gene Alias A030002D08Rik; F-BAR and double SH3 domains protein 1; FCH and double SH3 domains 1; FCH and double SH3 domains protein 1; Fchsd1; FLJ00007; nervous wreck homolog 2; NWK2; Protein nervous wreck 2; UNQ737/PRO1431
Gene Symbols FCHSD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FCHSD1. Recombinant protein control fragment (Product #RP-100294).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89848
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.