Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCHSD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558432
Description
Immunogen sequence: VEELSASENG DTPWMREIQI SPSPKPHASL PPLPLYDQPP SSPYPSPDKR SSLYFPRSPS ANEKSLHAES PGFSQASRHT PETSYGKLRP VRA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.
Adapter protein that plays a role in endocytosis via clathrin-coated pits. Contributes to the internalization of cell surface receptors, such as integrin ITGB1 and transferrin receptor (PubMed:29887380). Promotes endocytosis of EGFR in cancer cells, and thereby contributes to the down-regulation of EGFR signaling (PubMed:30249660). Recruited to clathrin-coated pits during a mid-to-late stage of assembly, where it is required for normal progress from U-shaped intermediate stage pits to terminal, omega-shaped pits (PubMed:29887380). Binds to membranes enriched in phosphatidylinositol 3,4-bisphosphate or phosphatidylinositol 3,4,5-trisphosphate (PubMed:29887380). When bound to membranes, promotes actin polymerization via its interaction with WAS and/or WASL which leads to the activation of the Arp2/3 complex. Does not promote actin polymerisation in the absence of membranes (PubMed:29887380). [UniProt].
Specifications
FCHSD2 | |
Polyclonal | |
Unconjugated | |
FCHSD2 | |
Carom; F-BAR and double SH3 domains protein 2; FCH and double SH3 domains 2; FCH and double SH3 domains protein 2; FCHSD2; KIAA0769; nervous wreck homolog; NWK; NWK1; Protein nervous wreck 1; SH3 multiple domains 3; SH3 multiple domains protein 3; SH3MD3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94868 | |
FCHSD2 | |
Recombinant protein corresponding to Human FCHSD2. Recombinant protein control fragment (Product #RP-102835). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction