Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131472
Description
FCP1 Polyclonal specifically detects FCP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FCP1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
CTDP1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human FCP1. The exact sequence is proprietary. | |
100 μL | |
Protein Phosphatase | |
9150 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
CCFDN, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) phosphatase, CTD of POLR2A, phosphatase of, subunit 1, EC 3.1.3.16, FCP1TFIIF-associating CTD phosphatase 1, RNA polymerase II subunit A C-terminal domain phosphatase, serine phosphatase FCP1a, subunit 1, TFIIF-associating CTD phosphatase, transcription factor IIF-associating CTD phosphatase 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction