Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559158
Description
Immunogen sequence: HPVVMKGLCA ECGQDLTQLQ SKNGKQQVPL STATVSMVHS VPELMVSSEQ AEQLGREDQQ RLHRNRKLVL MVDLDQTLIH TTEQHCQQMS NK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.
Specifications
FCP1 | |
Polyclonal | |
Unconjugated | |
CTDP1 | |
4930563P03Rik; AW553592; CCFDN; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) phosphatase, subunit 1; CTD (carboxy-terminal domina, RNA polymerase II, polypeptide A) phosphatase, subunit 1; CTD of POLR2A, phosphatase of, subunit 1; CTD phosphatase subunit 1; Ctdp1; FCP1; RNA polymerase II subunit A C-terminal domain phosphatase; serine phosphatase FCP1a; TFIIF-associating CTD phosphatase; TFIIF-associating CTD phosphatase 1; transcription factor IIF-associating CTD phosphatase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5B0 | |
CTDP1 | |
Recombinant protein corresponding to Human FCP1. Recombinant protein control fragment (Product #RP-98238). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction