Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FcRH5, Mouse anti-Human, Clone: F25, Millipore Sigma™

Catalog No. MABF2103100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2103100 100 μg
MABF210325 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2103100 Supplier MilliporeSigma Supplier No. MABF2103100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FcRH5, clone F25, Cat. No. MABF2103, is a mouse monoclonal antibody that detects Fc receptor-like protein 5 and has been tested for use in ELISA, Flow Cytometry, and Western Blotting.

Fc receptor-like protein 5 (UniProt: Q96RD9; also known as FcR-like protein 5, FcRL5, BXMAS1, Fc receptor homolog 5, FcRH5, Immune receptor translocation-associated protein 2, IRTA2, CD307e) is encoded by the FCRL5 (also known as FCRH5, IRTA2, UNQ503/PRO820) gene (Gene ID: 83416) in human. FcRH5 is a single-pass type I membrane protein that is expressed in marginal zone B-cells, immunoblasts, tonsillar germinal center centrocytes and in the intraepithelial and interfollicular regions of the tonsil. It is involved in B-cell development and differentiation in peripheral lymphoid organs and may be useful markers of B-cell stages. It is shown to regulate BCR signaling and binds aggregated IgG and engages several of its Ig-domains to bind IgG. It is shown to be overexpressed in many lymphoma cell lines and on hairy cell leukemia cells. Serum levels of soluble FcRH5 are reported to be elevated in patients with several types of B cell tumors. FcRH5 is synthesized with a signal peptide (aa 1-15), which is cleaved off in the mature form that has a long extracellular domain (aa 16-851), a short transmembrane domain (aa 852-872), and a cytoplasmic tail (aa 873-977) that contains immunoreceptor tyrosine-based inhibitor motifs (ITIM). (Ref.: Franco, A., et al. (2013). J. Immunol. 190(11); 5739-46; Ise, T., et al. (2007). Leukemia 21(1); 169-174).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcRH5
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone F25
Conjugate Unconjugated
Dilution Flow Cytometry Analysis: A representative lot detected FcRH5 in Flow Cytometry applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).Western Blotting Analysis: A representative lot detected FcRH5 in Western Blotting applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).ELISA Analysis: A representative lot detected FcRH5 in ELISA applications (Ise, T., et. al. (2005). Clin Cancer Res. 11(1):87-96).
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Fc receptor-like protein 5;FcR-like protein 5;FcRL5;BXMAS1;Fc receptor homolog 5;Immune receptor translocation-associated protein 2;IRTA2;CD307e
Gene Symbols FCRL5;FCRH5;IRTA2;UNQ503/PRO820
Host Species Mouse
Immunogen Full-length FcRH5 expression plasmid followed by a boost with 293T cells transfected with FcRH5 plasmid.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_112571.2
Test Specificity Clone F25 detects human Fc receptor-like protein 5. It targets an epitope with in the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.