Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL1/FcRH1 Antibody (282415), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2049PECY7
Description
FCRL1/FcRH1 Monoclonal antibody specifically detects FCRL1/FcRH1 in Human samples. It is validated for Flow CytometrySpecifications
FCRL1/FcRH1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
115350 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
282415 | |
Flow Cytometry | |
CD307a antigen, Fc receptor homolog 1, Fc receptor-like 1, FcRH1, FCRH1DKFZp667O1421, FcRL1, FcR-like protein 1, hIFGP1, IFGP family protein 1, IFGP1Fc receptor-like protein 1, Immune receptor translocation-associated protein 5, IRTA5immunoglobulin superfamily Fc receptor, gp42 | |
Mouse myeloma cell line NS0-derived recombinant human FCRL1/FcRH1, Ala17-His304, Accession # Q96LA6 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction