Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581387 Supplier Invitrogen™ Supplier No. PA581387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse FCRL1/FCRH1.

Fc receptor-like 1 (FCRL1) is a gene situated on chromosome 1q23, encoding a protein that is part of the Fc receptor-like family, which shares structural similarities with classical Fc receptors involved in immune responses. FCRL1 is predominantly expressed in B lymphocytes and is thought to play a role in the regulation of immune functions, particularly those associated with B cell development and differentiation. Although its precise biological function is not fully understood, it is believed to act as an immunomodulatory receptor that could influence B cell signaling pathways. The expression patterns of FCRL1 suggest possible involvement in autoimmune disorders and immune system modulation, potentially contributing to pathological conditions when dysregulated. Research into FCRL1 and other Fc receptor-like proteins continues to explore their contributions to immune homeostasis and potential as therapeutic targets in diseases involving B cell dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Fcrl1
Gene Accession No. Q8R4Y0
Gene Alias A230020G22Rik; Bxmas1; BXMAS1-like protein 1; BXMAS1-like protein 1b; Bxmh1; Bxmh1b; CD307a; CD307a antigen; fc receptor 1-like protein; fc receptor homolog 1; Fc receptor like 1; Fc receptor-like 1; Fc receptor-like protein 1; Fcrh1; FCRH1DKFZp667O1421; Fcrh1l; Fcrh1s; Fcrl1; FcR-like protein 1; hIFGP1; IFGP family protein 1; IFGP1; immune receptor translocation-associated protein 5; immunoglobulin superfamily Fc receptor, gp42; IRTA5; mBXMH1; mIFGP1
Gene Symbols Fcrl1
Host Species Rabbit
Immunogen Recombinant Mouse FCRL1/FCRH1 protein (Met1-Thr204).
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229499
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.