Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143166
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143166 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143166 Supplier Invitrogen™ Supplier No. PA5143166
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform 1 (NP_443170.1) and isoform 3 (NP_001152870.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

Fc receptor-like 1 (FCRL1) is a gene situated on chromosome 1q23, encoding a protein that is part of the Fc receptor-like family, which shares structural similarities with classical Fc receptors involved in immune responses. FCRL1 is predominantly expressed in B lymphocytes and is thought to play a role in the regulation of immune functions, particularly those associated with B cell development and differentiation. Although its precise biological function is not fully understood, it is believed to act as an immunomodulatory receptor that could influence B cell signaling pathways. The expression patterns of FCRL1 suggest possible involvement in autoimmune disorders and immune system modulation, potentially contributing to pathological conditions when dysregulated. Research into FCRL1 and other Fc receptor-like proteins continues to explore their contributions to immune homeostasis and potential as therapeutic targets in diseases involving B cell dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Fcrl1
Gene Accession No. Q96LA6
Gene Alias A230020G22Rik; Bxmas1; BXMAS1-like protein 1; BXMAS1-like protein 1b; Bxmh1; Bxmh1b; CD307a; CD307a antigen; fc receptor 1-like protein; fc receptor homolog 1; Fc receptor like 1; Fc receptor-like 1; Fc receptor-like protein 1; Fcrh1; FCRH1DKFZp667O1421; Fcrh1l; Fcrh1s; Fcrl1; FcR-like protein 1; hIFGP1; IFGP family protein 1; IFGP1; immune receptor translocation-associated protein 5; immunoglobulin superfamily Fc receptor, gp42; IRTA5; mBXMH1; mIFGP1
Gene Symbols Fcrl1
Host Species Goat
Immunogen Peptide with sequence C-SRLRKANITDVD.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115350
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.