Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL2/FcRH2 Antibody (B24), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311832DL594
Description
FCRL2/FcRH2 Monoclonal antibody specifically detects FCRL2/FcRH2 in Human samples. It is validated for Flow CytometrySpecifications
FCRL2/FcRH2 | |
Monoclonal | |
DyLight 594 | |
CD307b antigen, Fc receptor homolog 2, Fc receptor-like 2, FcRH2, FCRH2SPAP1B, FcRL2, FcR-like protein 2, IFGP family protein 4, IFGP4, Immunoglobulin receptor translocation-associated protein 4, immunoglobulin superfamily Fc receptor, gp42, IRTA4SPAP1C, SH2 domain containing phosphatase anchor protein 1, SH2 domain-containing phosphatase anchor protein 1Fc receptor-like protein 2, SPAP1A, SPAP1immune receptor translocation-associated protein 4 | |
DNA-immunization followed by a boost with FCRL2/FcRH2 transfected cells | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
B24 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
79368 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction