Learn More
Invitrogen™ FCRL2/FCRL5 Monoclonal Antibody (296902)

Description
Detects human FCRL2/FcRH2 and FCRL5/FcRH5 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human FCRL1, FCRL3, and FCRL4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | FCRL2/FCRL5 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 296902 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | FCRL2 |
| Gene Accession No. | Q96LA5, Q96RD9 |
| Gene Alias | RP11-367J7.6, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.