Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143168
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143168 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143168 Supplier Invitrogen™ Supplier No. PA5143168
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

The FCRL2 gene encodes the protein Fc receptor-like 2, which is part of the Fc receptor-like family involved in immune system regulation. FCRL2 is primarily expressed on the surface of B lymphocytes, where it contributes to the modulation of B cell activation and immune responses. As a member of the Fc receptor-like family, FCRL2 is thought to play roles in signaling pathways that influence B cell development, differentiation, and antibody production. It may act as a regulatory receptor that balances immune activation and inhibition, thereby contributing to immune homeostasis and prevention of autoimmunity. The expression of FCRL2 can impact conditions such as autoimmune diseases and B cell-related disorders, where dysregulation of B cell signaling is evident. Research into FCRL2 is ongoing to explore its specific ligand interactions and functional mechanisms, aiming to understand its potential as a therapeutic target for modulating immune responses in diseases characterized by B cell dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FCRL2
Gene Accession No. Q96LA5
Gene Alias CD307b; Fc receptor homolog 2; Fc receptor like 2; Fc receptor-like 2; Fc receptor-like protein 2; FCRH2; FCRL2; FcR-like protein 2; IFGP family protein 4; IFGP4; immune receptor translocation-associated protein 4; immunoglobulin receptor translocation-associated protein 4; immunoglobulin superfamily Fc receptor, gp42; IRTA4; RP11-367J7.6; SH2 domain containing phosphatase anchor protein 1; SH2 domain-containing phosphatase anchor protein 1; SPAP1; SPAP1A; SPAP1B; SPAP1C; UNQ9236/PRO31998
Gene Symbols FCRL2
Host Species Goat
Immunogen Peptide with sequence C-DKTSNIVKIKVQE.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79368
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.