Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL3/FcRH3 Antibody (H5), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311831AF647
Description
FCRL3/FcRH3 Monoclonal antibody specifically detects FCRL3/FcRH3 in Human samples. It is validated for Flow CytometrySpecifications
FCRL3/FcRH3 | |
Monoclonal | |
Alexa Fluor 647 | |
CD307c, Fc receptor homolog 3, Fc receptor-like 3, Fc receptor-like protein 3, FCRH3, fcR-like protein 3, hIFGP3, IFGP3, immunoglobulin superfamily receptor translocation associated protein 3, IRTA3, SPAP2 | |
DNA-immunization followed by a boost with FCRL3/FcRH3 transfected cells | |
0.1 mL | |
B Cell Development and Differentiation Markers | |
115352 | |
Store at 4°C in the dark. | |
IgG2b κ |
Flow Cytometry | |
H5 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction