Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL3/FcRH3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309272100UL
Description
FCRL3/FcRH3 Polyclonal specifically detects FCRL3/FcRH3 in Human samples. It is validated for Western Blot.Specifications
FCRL3/FcRH3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
CD307c, Fc receptor homolog 3, Fc receptor-like 3, Fc receptor-like protein 3, FCRH3, fcR-like protein 3, hIFGP3, IFGP3, immunoglobulin superfamily receptor translocation associated protein 3, IRTA3, SPAP2 | |
The immunogen is a synthetic peptide directed towards the middle region of Human FCRL3/FcRH3. Peptide sequence RGSSLSDAVHVEFSPDWLILQALHPVFEGDNVILRCQGKDNKNTHQKVYY | |
100 μg | |
B Cell Development and Differentiation Markers | |
115352 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction