Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547453
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547453 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547453 Supplier Invitrogen™ Supplier No. PA547453
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) IRTA2 and rhIRTA5 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL3
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene FCRL3
Gene Accession No. Q96P31
Gene Alias BXMAS1-like protein 2; CD307c; CD307e; Fc receptor homolog 3; Fc receptor like 3; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRL3; Fcrl5; fcR-like protein 3; FcR-like protein 5; hIFGP3; IFGP family protein 3; IFGP3; Immune receptor translocation-associated protein 3; immunoglobulin superfamily receptor translocation associated protein 3; IRTA3; mBXMH2; moFcRH3; rCG62831-like; SH2 domain-containing phosphatase anchor protein 2; SPAP2
Gene Symbols FCRL3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human FCRL3/FcRH3 Arg14-Arg569.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115352
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.