Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587910 Supplier Invitrogen™ Supplier No. PA587910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVAPKAVLLL NPPWSTAFKG EKVALICSSI SHSLAQGDTY WYHDEKLLKI KHDKIQITEP GNYQCKTRGS SLSDAVHVEF SPDWLILQAL HPVFEGDNVI LRCQGKDNKN THQKVYYKDG KQLPNSYNLE KITVNSVSRD NSKYHCTAYR KFYILDIEVT SKP; Positive Samples: U-937, H460, 293T, HeLa, Mouse kidney; Cellular Location: Cell membrane, Single-pass type I membrane protein.

This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FCRL3
Gene Accession No. Q96P31
Gene Alias BXMAS1-like protein 2; CD307c; CD307e; Fc receptor homolog 3; Fc receptor like 3; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRL3; Fcrl5; fcR-like protein 3; FcR-like protein 5; hIFGP3; IFGP family protein 3; IFGP3; Immune receptor translocation-associated protein 3; immunoglobulin superfamily receptor translocation associated protein 3; IRTA3; mBXMH2; moFcRH3; rCG62831-like; SH2 domain-containing phosphatase anchor protein 2; SPAP2
Gene Symbols FCRL3
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115352
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.