Learn More
Invitrogen™ FCRL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120092
Description
Positive test controls include: Raji, B cells, 293T, U-937, Mouse kidney, Rat kidney. The target is usually found in the following locations: Cell membrane, Single-pass type I membrane protein. Immunogen sequence: RRKPGGLSAT GTSSHSPSEC QEPSSSRPSR IDPQEPTHSK PLAPMELEPM YSNVNPGDSN PIYSQIWSIQ HTKENSANCP MMHQEHEELT VLYSELKKTH PDDSAGEASS RGRAHEEDDE E.
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
Specifications
FCRL3 | |
Polyclonal | |
Unconjugated | |
FCRL3 | |
BXMAS1-like protein 2; CD307c; CD307e; Fc receptor homolog 3; Fc receptor like 3; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRL3; Fcrl5; fcR-like protein 3; FcR-like protein 5; hIFGP3; IFGP family protein 3; IFGP3; Immune receptor translocation-associated protein 3; immunoglobulin superfamily receptor translocation associated protein 3; IRTA3; mBXMH2; moFcRH3; rCG62831-like; SH2 domain-containing phosphatase anchor protein 2; SPAP2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
115352 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.92 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96P31 | |
FCRL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 600-720 of human FCRL3 (NP_443171.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.