Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL4/FcRH4/IRTA1 Antibody (A1), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311836UV
Description
FCRL4/FcRH4/IRTA1 Monoclonal antibody specifically detects FCRL4/FcRH4/IRTA1 in Human samples. It is validated for Flow CytometrySpecifications
FCRL4/FcRH4/IRTA1 | |
Monoclonal | |
DyLight 350 | |
CD307d antigen, Fc receptor homolog 4, Fc receptor-like 4, FcRH4, FCRH4Fc receptor-like protein 4, FcRL4, FcR-like protein 4, hIFGP2, IFGP family protein 2, IFGP2, IGFP2, Immune receptor translocation-associated protein 1, immunoglobulin superfamily Fc receptor, gp42, immunoglobulin superfamily receptor translocation associated 1, IRTA1MGC150523, MGC150522 | |
DNA-immunization followed by a boost with FCRL4/FcRH4/IRTA1-transfected cells | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
A1 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
83417 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction