Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCRL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598662
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598662 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598662 Supplier Invitrogen™ Supplier No. PA598662
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FCRL4 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein has four extracellular C2-type immunoglobulin domains, a transmembrane domain and a cytoplasmic domain that contains three immune-receptor tyrosine-based inhibitory motifs. This protein may play a role in the function of memory B-cells in the epithelia. Aberrations in the chromosomal region encoding this gene are associated with non-Hodgkin lymphoma and multiple myeloma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FCRL4
Gene Accession No. Q96PJ5
Gene Alias CD307d; fc receptor homolog 4; Fc receptor like 4; Fc receptor-like 4; Fc receptor-like protein 4; FcRH4; FCRL4; FcR-like protein 4; hIFGP2; IFGP family protein 2; IFGP2; IGFP2; immune receptor translocation-associated protein 1; immunoglobulin superfamily Fc receptor, gp42; immunoglobulin superfamily receptor translocation associated 1; IRTA1; MGC150522; MGC150523
Gene Symbols FCRL4
Host Species Rabbit
Immunogen Recombinant Human Fc receptor-like protein 4 protein (20-387AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83417
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.