Learn More
Invitrogen™ FCRL4 Polyclonal Antibody

Description
PA1-4233 detects the IRTA1 protein in human samples. PA1-4233 has been successfully used in Western blot procedures. PA1-4233 immunizing peptide corresponds to amino acid residues 69-84 of the human IRTA1 protein.
Specifications
Specifications
| Antigen | FCRL4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FCRL4 |
| Gene Accession No. | Q96PJ5 |
| Gene Alias | CD307d; fc receptor homolog 4; Fc receptor like 4; Fc receptor-like 4; Fc receptor-like protein 4; FcRH4; FCRL4; FcR-like protein 4; hIFGP2; IFGP family protein 2; IFGP2; IGFP2; immune receptor translocation-associated protein 1; immunoglobulin superfamily Fc receptor, gp42; immunoglobulin superfamily receptor translocation associated 1; IRTA1; MGC150522; MGC150523 |
| Gene Symbols | FCRL4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.