Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL5 (CD307e) Mouse anti-Human, PE, Clone: 509F6, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566734
Description
FCRL5 (CD307e) Monoclonal antibody specifically detects FCRL5 (CD307e) in Human samples. It is validated for Flow Cytometry.
Specifications
FCRL5 (CD307e) | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
FCRL5 (CD307e) | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
509F6 | |
PE | |
50 μg | |
Primary | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction