Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRL5 (CD307e) Mouse, anti-Human, R718, Clone: 509F6, BD OptiBuild™

Catalog No. BDB751885 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751885 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751885 Supplier BD Biosciences Supplier No. 751885
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 509F6 monoclonal antibody specifically binds to Fc receptor-like 5 (FCLR5), a type I transmembrane glycoprotein that is also known as CD307e, FCRH5 (Fc receptor homolog 5) and IRTA2 (immunoglobulin superfamily receptor translocation associated 2). FCRL5 (CD307e) is a member of the Immunoglobulin (Ig) gene superfamily with 9 extracellular Ig domains, an intracellular immunoreceptor tyrosine-based activating motif (ITAM)-like sequence and two intracellular immunoreceptor tyrosine-based inhibitory motif (ITIM)-like sequences. Like other Fc receptor-like proteins, FCRL5 (CD307e) is homologous to FcγR1 (CD64) and binds to immunoglobulins, specifically the IgG isotypes, and it may play a role in the regulatory interactions of immune complexes with B lymphocytes. FCRL5 (CD307e) is differentially expressed in the B cell lineage, with little expression on peripheral blood cells and upregulated expression on plasma cells and some leukemias and lymphomas. mAb 509F6 binds to an epitope within the first 3 Ig domains of FCRL5 and blocks FCRL5's interaction with immune complexes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL5 (CD307e)
Applications Flow Cytometry
Classification Monoclonal
Clone 509F6
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.